Ultrasone vloeistofprocessors, programmeerbaar, Sonicator®
Leverancier: QSONICA LLC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Q700-220 Q500A-220 Q500-220 Q700A-220 Q500AUK-220 Q500UK-220 Q700AUK-220
432-0237EA 6190 EUR
432-0237 432-0234 432-0233 432-0238 QSONQ500AUK-220 QSONQ500UK-220 QSONQ700AUK-220
Ultrasone vloeistofprocessors, programmeerbaar, Sonicator®
Homogenisatoren Ultrasone celscheiders
A range of programmable ultrasonic processors with applications including cell lysis, nanoparticle dispersion, creating emulsions and general homogenisation. Pulses with 20 KHz frequency over a range of 500 to 700 W depending on the model.
  • Q500:
  • Programmable operation by setting time and amplitude
  • Pulse mode prevents heat build up in temperature sensitive samples
  • Digital amplitude/intensity control
  • Real time energy monitoring
The Q700 introduces some additional features designed to ensure reproducibility of results and flexibility of operation.
Full amplitude control 1 to 100% for identifying optimum settings for efficient sample processing
Programmability to run multiple programs in sequence
Auto-tuning to maintain electrical efficiency with changes in load and temperature
Touch screen interface offers intuitive control
The Q700 introduces some additional features designed to ensure reproducibility of results and flexibility of operation.
Leveringsinformatie: sonicatorpakket omvat generator, omvormer, stroomkabel, omvormerkabel, sleutelset en sonde met een diameter van ¹/₂" inch.
Opgelet: gebruik nooit een sonde met vervangtips bij vloeistoffen met lage oppervlaktespanning. Gebruik daarvoor een sonde met vaste tip.