Large Rectangular Waste Bins
Leverancier: Gilac PROFESSIONNEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G140521 G145521 G145621 G145821
216-2880EA 384 EUR
216-2880 CORAG145521 CORAG145621 CORAG145821
Large Rectangular Waste Bins
Bakken Afvalbakken
LDPE.
  • Mechanical and chemical resistance
  • Durability and versatility
  • Easy to clean