Antibody saver trays, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F451000000 F451000001 F451000002
BELAF451000000EA 87.2 EUR
BELAF451000000 BELAF451000001 BELAF451000002
Antibody saver trays, SP Bel-Art
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
Antibody saver trays, PP, translucent, minimizes the use of expensive antibodies when incubating filters in the western blot process by using an optimally sized tray.
  • Single lane tray in a set of five linked together for easier handling but can be easily separated
  • Available for popular midi and mini gels
  • Includes attached lid to protect against spillage
  • Trays are stackable, enclosable and reusable