Adhesion slides for capillary gap stainers, HistoBond®
Leverancier: Marienfeld
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HistoBond®
MARI0890004EA 0 EUR
MARI0890004
Adhesion slides for capillary gap stainers, HistoBond®
Object- en dekglaasjes Adhesie-objectglaasjes
Adhesion slides are specially designed to endure harsh procedures.
  • Pre-cleaned, ready for use
  • Steps repeated until the slides tissue sections have been fully exposed to all required reagents
  • 80 μm high spacer stripe and a raised frosted end
Used with staining equipment for immunochemistry.
Certificaten: ISO 8037/1
Verpakking: Boxes of 50 pieces − 50 boxes per carton.