Micro pipettes, disposable, calibrated
Leverancier: DRUMMOND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2-000-001 2-000-005 2-000-010 2-000-020 2-000-025 2-000-030 2-000-040 2-000-044 2-000-050 2-000-200 2-000-201 2-000-100 2-000-075
DRUM2-000-001EA 146 EUR
DRUM2-000-001 DRUM2-000-005 DRUM2-000-010 DRUM2-000-020 DRUM2-000-025 DRUM2-000-030 DRUM2-000-040 DRUM2-000-044 DRUM2-000-050 DRUM2-000-200 DRUM2-000-201 DRUM2-000-100 DRUM2-000-075
Micro pipettes, disposable, calibrated
Pipetten Micro-capillairen
These precision calibrated micro pipettes are available in sizes ranging from 1 to 200 µl. Ideal for a variety of uses in liquid transfer work.
  • Fire polished
  • Accuracy of ±0,5%
  • Colour coded micro pipettes with sizes imprinted
  • Anticoagulant treatment for clinical applications (special limited sizes)
Verpakking: 250 colour coded micro pipettes, plus one 15" aspirator tube, are supplied in a conveniently designed, easy to store cylinder.