Hexnut fittings
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0730014 1021003 1021011
SGEA0730014EA 185 EUR
SGEA0730014 SGEA1021003 SGEA1021011
Hexnut fittings
Fittingen en connectoren
Stainless steel 10-32 thread fittings use a non-swaging PEEK replaceable ferrule, for use with GLT™ columns.
  • Inert contact surfaces make them biocompatible
  • Ideal for applications where corrosive solvents are being used
  • Maximum pressure rating of 344,74 bar (5000 psi)