Tubing cutter, Terry-Tool
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
082780 082782
SGEA082780EA 407 EUR
SGEA082780 SGEA082782
Tubing cutter, Terry-Tool
Snijders
Cutter for steel tubing.
  • Cutting wheel for cutting ¹/₁₆ and ¹/₈" stainless steel tubing
  • Clean, right angle cuts with minimal burring or chipping of the tubing
  • No additional reaming or deburring is required
Bestelinformatie: Spare cutting wheels are available.