Cation exchange HPLC columns, Dionex™, IonPac™ CS12A
Leverancier: DIONEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
046074 046073
DION046074EA 0 EUR
DION046074 DION046073
Cation exchange HPLC columns, Dionex™, IonPac™ CS12A
Kolommen voor chromatografie
CS12A columns are recommended for the fast isocratic separation of lithium, sodium, ammonium, potassium, magnesium and calcium from the sample matrices.
  • Compatible with organic solvents
  • High temperature
  • High acid concentrations
Sample matrices are like environmental and drinking water, soil extracts, acid digest and power plant waters treated with ammonium or morpholine, and also industrial solvents.