Natriumchloride Water_softening
Leverancier: SMEG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Saline, NaCl, Sea salt, Halite
980103 SALE
SMEG980103EA 51.1 EUR
SMEG980103 SMEGSALE
Natriumchloride Water_softening
Natriumchloride
Formule: NaCl
MW: 58.44 g/mol
Kookpunt: 1413
Smeltpunt: 801
MDL Nummer: MFCD00003477
CAS nummer: 7647-14-5
Merck Index: 14,08599