VWR® HistoChoice®, Clearing agent
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H103-4LEA 221 EUR
H103-4L H103-1L
VWR® HistoChoice®, Clearing agent
Kleurstoffen Microscopy and staining reagents
HistoChoice® clearing agent is a much less hazardous replacement for xylene/toluene.
  • Faster wax dissolution
  • Slower evaporation than xylene with no oily residue
  • Compatible with organic mounting media
Designed to work with isopropanol in automated processors.