Dissection boards for the food industry, Euroboard
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7056 7059 7061
SEMA7056EA 234 EUR
SEMA7056 SEMA7059 SEMA7061
Dissection boards for the food industry, Euroboard
Borden voor dissectie
These chopping boards are made from professional-grade HYGENIA® HDPE 500 material. Ideal for the food-service and food-processing industries.
  • In accordance with the international hygiene regulations and HACCP
  • No bacterial cross-contamination with other food groups
  • No transfer of tastes or smells
  • Unbreakable, easy to clean (dishwasher safe)
  • Protects knives, preserving their cutting ability
  • Equipped with small, non-slip feet
Different colours ensure clear assignment to different areas for maximum safety in your kitchen.
Colour assignment: red: meat /More Product Information