Measuring beakers with handle, PP
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4731 4732
SEMA4731EA 27.6 EUR
SEMA4731 SEMA4732
Measuring beakers with handle, PP
Bekers
PP, highly translucent.
  • Volume is equally visible for left and right handers
  • Good chemical resistance, resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
  • To preserve markings, cleaning at no higher than 60 °C is recommended
  • For autoclaving we recommend the design with raised graduations
Ideal for general laboratory use and a wide variety of other applications. With stable handle, spout and easily readable, printed black scale on both sides. Stackable.
Food-safe product according to EC Directive No. 10/2011