Test tubes, disposable
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1594 2168
SEMA1594EA 70.2 EUR
SEMA1594 SEMA2168
Test tubes, disposable
Buisjes Reageerbuisjes
PS, clear.
  • Good thermal resistance
  • Good chemical resistance
  • Robust, economical
Universal tubes for a wide range of applications.