Anion exchange HPLC columns, ProPac™ PA1
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
039657 039658 040137
555-7260EA 0 EUR
555-7260 555-7261 555-7262
Anion exchange HPLC columns, ProPac™ PA1
Kolommen voor chromatografie
Columns are designed for the separation and purification of hydrophilic anionic proteins and peptides.
  • Pellicular packing ensure high efficiency and fast mass transport
  • Ideal for high resolution separations