VWR®, DNA loading buffers
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
733-2576EA 44 EUR
733-2576 733-2575 733-2574
VWR®, DNA loading buffers
Kleurstoffen Kleuringen voor elektroforese
These DNA loading dyes are used to load DNA samples to agarose or SDS DNA gels for gel electrophoresis.
Suitable for TAE, TBE, SB and LB electrophoresis buffers.
Certificaten: EC Regulation No. 1907/2006 (REACH).
Verpakking: Each kit contains 5×1 ml.
Opgelet: Store at –20 °C for in vitro use only.