WetGrip labels
Leverancier: DIVERSIFIED BIOTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GRDT-2000 GRDT-1000 GRDT-3000 GRDT-4000 GRDT-6000 GRLA-2016 GRLA-1000 GRLA-2380 GRLA-1050
DIVEGRDT-2000EA 149 EUR
DIVEGRDT-2000 DIVEGRDT-1000 DIVEGRDT-3000 DIVEGRDT-4000 DIVEGRDT-6000 DIVEGRLA-2016 DIVEGRLA-1000 DIVEGRLA-2380 DIVEGRLA-1050
WetGrip labels
Etiketten
These WetGrip labels grip to moist, frosty or frozen surfaces.
  • Temperature resistant to −80 °C
  • Compatible with centrifuge rotors
  • Resistant to chemicals and solvents
  • Adheres to plastics, glass, metals and even paperboard boxes
Available in white laser sheets, white direct thermal rolls, or white hand writable rolls.