Luer-lock connectors, miniature, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F718-02 F718-04 F718-06 F718-08
BOHLF718-02EA 33 EUR
BOHLF718-02 BOHLF718-04 BOHLF718-06 BOHLF718-08
Luer-lock connectors, miniature, BOLA
Fittingen en connectoren
PTFE, white.
  • Temperature resistance from −200 to +250 °C
  • Universal chemical resistance
  • FDA conform
Connector for transition from Luer Lock connection to female thread UNF 1/4" 28G. The inner core of the Luer-Lock connection provides good sealing, the additional thread (either male or female) prevents accidental loosening. The tubing is connected by means of tube end fittings to a female thread UNF 1/4“ 28G. Connection of components with Luer Lock such as syringes or cannulas to a tubing system.