Magnetic stirrer heads with flat flange, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C522-40 C522-50
BOHLC522-40EA 1520 EUR
BOHLC522-40 BOHLC522-50
Magnetic stirrer heads with flat flange, BOLA
Roerkoppen
Magnetic stirrer heads consist of perfect capsulated drive shaft with ball bearings, rotor and lower bearing made of PTFE and a conductor made of borosilicate glass. These stirrer heads are designed with flat flanges sealed with gasket.
  • No grease required and safe to run dry
  • Height adjustable stirrer shaft (40 mm)
  • Long durability and compact size
  • No leakage or memory effects due to non-porous, welded rotor
  • Temperature range: −100 to 240 °C
  • Chemical resistance
  • Powerful transmission of upto 90 Ncm