Disposable cuvettes
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
67.758 67.758.001
SARS67.758EA 129.8 EUR
SARS67.758 SARS67.758.001
Disposable cuvettes
Cuvetten Cuvetten voor spectrofotometers
UV transparent and disposable cuvettes designed for photometric analyses.
Certified DNA-, RNase- and protein-free.
Verpakking: In a Styrofoam box.