HPLC column filters, direct-connection, Javelin™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88200 88400 88700
HYPE88200EA 250 EUR
HYPE88200 HYPE88400 HYPE88700
HPLC column filters, direct-connection, Javelin™
Filters Voorkolom filters en frits
Protect HPLC systems with one-piece Javelin™ direct-connection column filters.
  • Replace entire disposable filter unit for easy changes
  • Recommended for use as dedicated filters for a column rather than the HPLC system
  • CPI tip attaches directly to HPLC column inlet without tubing or wrenches
Order Now
Specificaties
Environmentally Preferable