SMD tweezers, Erem®
Leverancier: Basan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
102ACA 102ACAX
BSNC102ACAEA 55.2 EUR
BSNC102ACA BSNC102ACAX
SMD tweezers, Erem®
Pincetten
Stainless steel. These special tweezers are ideal for handling chips or miniature SMD components. Suitable for mounting on SMD boards or ceramic substrates.
  • With anti-glare finish, non magnetic
  • With angled head and blunt edges to avoid board damage