SMD tweezers, Erem®
Leverancier: Basan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
124791 150SAD T150SA
BSNC124791EA 40.6 EUR
BSNC124791 BSNC150SAD BSNCT150SA
SMD tweezers, Erem®
Pincetten
Stainless steel. These tweezers are ideal for the gripping and holding of cylindrical components. Available in straight and angled versions.
  • Non rusting, heat resistant, and anti-acid proof surface
  • Anti-magnetic
  • With blunted edges to prevent damage to printed-circuit boards