Capillary columns, traditional phases, SPB-5, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
25343 25347 25351 24101-U 24166 25304 25305-U 25317 25342 25350 25420-U 24034 24035 24036 24037 24048 24049 24050 24051
25343.EA 850 EUR
25343. 25347. 25351. 24101-U 24166. 25304. 25305-U 25317. 25342. 25350. 25420-U 24034. 24035. 24036. 24037. 24048. 24049. 24050. 24051.
Capillary columns, traditional phases, SPB-5, Supelco®
Kolommen voor chromatografie
These bonded poly(5% diphenyl/95% dimethylsiloxane) low phenyl content columns improve thermal stability of the phase, while still providing essentially a boiling point elution order, and a slight increase in selectivity, especially for aromatic compounds.
  • Temperature limit from –60 to 320 °C
Certificaten: Meets USP G27 and G36 requirements.