Pipette tips, Ovation®
Leverancier: VISTALAB TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4058-1000 4060-1002 4058-2000 4060-2132 4058-3000 4060-9026 4060-9024
613-4927EA 110 EUR
613-4927 613-4928 613-4933 613-4938 613-4940 VIST4060-9026 VIST4060-9024
Pipette tips, Ovation®
Pipettips
Ideal for use with Ovation and MLA pipettes.
  • Made of 100% pure virgin PP
  • Provide a perfect fit
  • Deliver reliable performance
  • Autoclavable