Air sampler, Spin Air
Leverancier: IUL Instruments SA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5502 5500
710-0357EA 2470 EUR
710-0357 710-0356
Air sampler, Spin Air
Monsternemers Luchtmonsternametoestellen
All the components has been made for long durability. Special design of the holes combined with slow rotation of the plate allow to use 100% of the surface compared with the 5% of the still devices.
  • Bar code connection
  • Connectivity to 2nd unit
  • Hard casing carrying moulded box
In order to cut the sampling time by two, just add one Slave connected to the main unit. The microprocessor and the power supply of the last one, will control the Slave. As final result two Petri dishes have been inoculated. In the back of the instrument there is a line of connections: Switching adapter for AC mains to recharge the internal batteries, connection Spin Air-Slave, bar-code reader, serial connection to PC/printer.
Leveringsinformatie: Supplied with 90 mm plate holder and cover from aluminium.