Pipette tips, universal
Leverancier: BOETTGER PAUL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
03.013.0103 03.023.0101 03.033.0100
BOET03.013.0103EA 16.1 EUR
BOET03.013.0103 BOET03.023.0101 BOET03.033.0100
Pipette tips, universal
Pipettips
These tips are made from selected, hydrophobic PP.
  • Optimal surface quality
  • Precise and centric opening of tip
  • For optimal use on single and multi-channel pipettes
  • Autoclavable at 121 °C/2,1 bar
  • High accuracy