Empty vials, depyrogenated
Leverancier: EP SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ST5-20 ST10-20 ST50-20 ST100-20 ST2-13 ST30-20 ST100-20A ST10-20A ST30-20A ST50-20A ST5-20A
EPSCST5-20EA 414 EUR
EPSCST5-20 EPSCST10-20 EPSCST50-20 EPSCST100-20 EPSCST2-13 EPSCST30-20 EPSCST100-20A EPSCST10-20A EPSCST30-20A EPSCST50-20A EPSCST5-20A
Empty vials, depyrogenated
flesjes
Clear depyrogenated sterile empty vials are type I borosilicate, assembled with butyl stoppers and aluminium seals. Used in the aseptic protocol.
  • Capacities ranges from 1 to 100 ml
  • Includes certificates of sterility and pyrogen test