Capillary columns, traditional phases, SPB-1, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
24028 25314 24213-U 24215-U 24230-U 24296 24297 25303 25341-U 25344 25345-U 25349 25360 25388 25417 24029 24030-U 24031 24044 24045-U 24047
24028.EA 757 EUR
24028. 25314. 24213-U 24215-U 24230-U 24296. 24297. 25303. 25341-U 25344. 25345-U 25349. 25360. 25388. 25417. 24029. 24030-U 24031. 24044. 24045-U 24047.
Capillary columns, traditional phases, SPB-1, Supelco®
Kolommen voor chromatografie
These non-polar methyl silicone bonded poly(dimethyl siloxane) columns are used to separate the sample components according to boiling point.
  • Bonded polymer matches the polarity of its nonbonded predecessors
  • Temperature limit from −60 to 320 °C
Certificaten: Meets USP G1, G2 and G9 requirements.