Capillary columns, traditional phases, SPB-624, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
24251 24255 24256 25430 28662-U
24251.EA 1310 EUR
24251. 24255. 24256. 25430. 28662-U
Capillary columns, traditional phases, SPB-624, Supelco®
Kolommen voor chromatografie
This column is specially designed for purge-and-trap analyses of volatile halogenated, non-halogenated, and aromatic contaminants from environmental samples.
  • Bonded, low bleed
  • Proprietary
  • High thermal stability - maximum temperatures up to 250 °C