Capillary columns, traditional phases, VOCOL, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
24157 24154 24205-U 24217-U 25320-U 25358 25381 28463-U
24157.EA 1490 EUR
24157. 24154. 24205-U 24217-U 25320-U 25358. 25381. 28463-U
Capillary columns, traditional phases, VOCOL, Supelco®
Kolommen voor chromatografie
This intermediate polarity column is mainly designed for analyses of volatile organic compounds (VOCs), offers great retention and resolution of highly volatile compounds.
  • Bonded, proprietary
  • High thermal stability - maximum temperatures up to 250 °C