Koudebestendige handschoenen, Arctic Winter Glove, A146
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A146YERXL A146YERL A146YERXXL
PRTWA146YERXLEA 8.6 EUR
PRTWA146YERXL PRTWA146YERL PRTWA146YERXXL
Koudebestendige handschoenen, Arctic Winter Glove, A146
Handschoenen
Deze nylon handschoenen met nitril coating bieden bescherming tegen koude en mechanische gevaren.
  • Twin liner traps in heat with a ¾ dipped coating to keep the hands dry
  • Soft nitrile foam dipping offers full protection to the fingers
  • Flexible sandy nitrile coating offers great grip in wet and dry conditions
  • Open back for breathability
  • Superb abrasion and tear resistance
  • Low linting construction for minimal contamination
Specially designed for use in cold conditions.
Certificaten: EN 388:2016 (4242X), EN 407 (X2XXXX), EN 511 (X2X).