Gas sampling bags, Tedlar®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
24633 24634 24654 24655
24633.EA 167 EUR
24633. 24634. 24654. 24655.
Gas sampling bags, Tedlar®
Zakken Gaszakken
The unique valve and seam-sealing process ensure that these bags are sturdy and virtually leak-proof, even under the most demanding sampling and shipping conditions.
  • Valve consistently achieves a positive open or closed setting
  • 2 mil DuPont Tedlar material is used for superior inertness and impermeability
  • Patented polypropylene 2-in-1 valve has a unique push-pull function which allows the operating of bag’s valve effectively with one hand, while preparing a connection or withdrawing an aliquot with the other hand