Level control systems with signalboxes, SymLine
Leverancier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
117988 108157 108125 108088 108087 108158
SCTV117988EA 1790 EUR
SCTV117988 SCTV108157 SCTV108125 SCTV108088 SCTV108087 SCTV108158
Level control systems with signalboxes, SymLine
Niveausensoren
Monitor two filling levels at once, perfect integration into lab furniture with the Built-In Signalbox ant the Signalbox T1 and T4.
  • For all types of Safety Waste Caps and other electronic level controllers
  • Touchless alarm-resetting fields for two containers
  • 2 switch inputs and 2 switch outputs for peripheral units
LEDs and acoustic buzzer warn you in due time before the filling level of your collection containers reaches a critical height. When the container is changed, the alert can be muted via the new touchpad. Pumps and other external devices can be conveniently triggered via the integrated interface.