Analytical HPLC columns, Symmetry® C18
Leverancier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WAT054275 WAT046980 WAT066224 WAT200632 WAT045905 WAT054225 186000695 WAT056975 186000690 186002090 WAT066220 WAT200625 186000207
WATEWAT054275EA 0 EUR
WATEWAT054275 WATEWAT046980 WATEWAT066224 WATEWAT200632 WATEWAT045905 WATEWAT054225 WATE186000695 WATEWAT056975 WATE186000690 WATE186002090 WATEWAT066220 WATEWAT200625 WATE186000207
Analytical HPLC columns, Symmetry® C18
Kolommen voor chromatografie
Symmetry® C18 columns feature monofunctionally bonded C18 ligands on a high purity base-deactivated silica.
Symmetry® columns have the tightest column specifications in the industry giving resulting highly reproducible chromatographic results.