Reversed phase HPLC columns, Atlantis® dC18
Leverancier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
186001342 186001299 186001346 186001287 186001337 186001329 186001333 186001323
WATE186001342EA 970 EUR
WATE186001342 WATE186001299 WATE186001346 WATE186001287 WATE186001337 WATE186001329 WATE186001333 WATE186001323
Reversed phase HPLC columns, Atlantis® dC18
Kolommen voor chromatografie
Atlantis® dC18 columns are universal, silica-based, reversed phase C18 columns that are used for polar compound retention. The Atlantis® dC18 columns feature di-functionally bonded C18 ligands that have been optimised for use with highly aqueous mobile phases, including 100% water.