Tool cart
Leverancier: Palbam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PALBCT0702EA 5240 EUR
PALBCT0702
Tool cart
Transportwagens
Tool cart in stainless steel 304 with electropolished finish.
  • Five fully extendable drawers
  • One lock which secures all the drawers
  • Top ledge to prevent tools/parts falling
  • Cleanroom castors with brakes