Pass through cabinets
Leverancier: Palbam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PT-604060 PT-706070MS
PALBPT-604060EA 0 EUR
PALBPT-604060 PALBPT-706070MS
Pass through cabinets
Meubilair Opslagkasten
Pass Through cabinet allows products and materials to be transferred between different rooms. Reduces unnecessary walking and entering/exiting cleanroom.
  • Single or double versions available
  • Sealing gaskets and compression handles ensure firm closure
  • Tempered Glass or anti static PVC windows available
  • Mechanical or electronic interlocks available
  • Can be customised to requirements
Fabrication material: 304 stainless steel with electropolished finish.