Wipe down table
Leverancier: Palbam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WDT-1260 WDT-0960
134-0489EA 0 EUR
134-0489 132-1091
Wipe down table
Meubilair Tafels
The wipe down table is designed to support the wipe down procedure. Includes a hole in the table top for easy waste disposal.
The cabinet under the table and under the waste disposal hole has a removable plastic bin included inside.
In stainless steel 304 with electropolished finish.