HPTLC plates, nano silica gel 60, unmodified layers, with concentrating zone, Nano-SILGUR
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
552-0231EA 255 EUR
552-0231 552-0232 MANA818442 MANA818432
HPTLC plates, nano silica gel 60, unmodified layers, with concentrating zone, Nano-SILGUR
TLC-platen
Unmodified HPTLC silica layers with concentrating zone for HPTLC.
  • Nano silica gel (particle size 2 to 10 μm) with kieselguhr zone
  • Rapid sample application
  • Always regular substance spots
Nano-SILGUR layers combine the advantages of HPTLC (sharper separations, shorter developing times, increased detection sensitivity) and of the concentrating zone (regular spots, reliable detection).
Nano silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 2 to 10 μm.
Kieselguhr: Small surface of low activity.