Overalls, Sealtex™ Classic S452
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S452NARXXL S452NARS S452NARM S452NARL S452NARXXXL S452NARXL
PRTWS452NARXXLEA 91.9 EUR
PRTWS452NARXXL PRTWS452NARS PRTWS452NARM PRTWS452NARL PRTWS452NARXXXL PRTWS452NARXL
Overalls, Sealtex™ Classic S452
Werkkleding
Complete waterproof overall made of Sealtex™ Classic, a high technology PU coated fabric on a flexible, highly resistant polyester tricot knit. The stretchable fabric is tear resistant, anti-fungal, oil, fat, diesel and grease resistant and can be easily wiped clean.
  • Zip front fastening with studded storm flap which prevents water from penetrating
  • Side access pockets
  • Roll-away hood with drawcord
  • Elasticated back waist
  • Finished with high frequency welded seams ensuring maximum water resistance
Colour: Navy blue
Weight: 200 g/m²
Certificaten: EN 343 (3-1)