High visibility bomber jacket, S463
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S463YERXL S463YER5XL S463YERXXL S463YERL S463YERXS S463YER6XL S463YERS S463YERM S463YERXXXL S463YER4XL S463YER7XL S463ORR4XL S463ORR5XL S463ORRL S463ORRM S463ORRS S463ORRXL S463ORRXS S463ORRXXL S463ORRXXXL
PRTWS463YERXLEA 45 EUR
PRTWS463YERXL PRTWS463YER5XL PRTWS463YERXXL PRTWS463YERL PRTWS463YERXS PRTWS463YER6XL PRTWS463YERS PRTWS463YERM PRTWS463YERXXXL PRTWS463YER4XL PRTWS463YER7XL PRTWS463ORR4XL PRTWS463ORR5XL PRTWS463ORRL PRTWS463ORRM PRTWS463ORRS PRTWS463ORRXL PRTWS463ORRXS PRTWS463ORRXXL PRTWS463ORRXXXL
High visibility bomber jacket, S463
Werkkleding
This jacket is made of 300D Oxford weave polyester with a stain resistant finish and PU coating. Completely padded and lined, the jacket offers great freedom of movement.
  • Two hip pockets with Velcro fastened flaps
  • Two-way zip front with studded storm flap, drawstring hood
  • Knitted storm cuffs, taped waterproof seams
  • Inside pocket, mobile phone pocket
Weight: 190 g/m²
Weight: 190 g/m²
Certificaten: EN 343 (3-1), EN 342 (Class 3), EN ISO 20471 (Class 3)