Erlenmeyer flasks, baffled, Delong neck
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1141/08 1141/10 1141/12 1141/14
214-0431EA 883 EUR
214-0431 214-0432 214-0433 214-0434
Erlenmeyer flasks, baffled, Delong neck
Kolven Erlenmeyers
PYREX®, borosilicate glass.
  • Four equidistant baffles provide consistent agitation when used with compatible orbital shakers
  • Excellent chemical resistance, high temperature resistance
  • Flasks are supplied without closures
Ideal for aerobic microbiology and cell culture.