Instrument trays, with lid
Leverancier: Agencinox
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5029 5036 5046 5052 5075
AGEN5029EA 0 EUR
AGEN5029 AGEN5036 AGEN5046 AGEN5052 AGEN5075
Instrument trays, with lid
Reservoirs/Schalen/Potten Instrumentenschalen
Stainless steel, non-rusting.
  • Special corner shape for fast, easy cleaning
  • Polished electrolytically, anti-magnetic
  • Good chemical resistance
  • Excellent thermal resistance
With a cover that enables the instrument dishes to be stacked.