GC septa, Thermogreen® LB-1
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20657-U 20658 20661
20657-UEA 0 EUR
20657-U SUPL20658 20661.
GC septa, Thermogreen® LB-1
Tussenschoten
These septa are suitable for use with 50 to 300 °C inlet temperatures.