Safety wash bottles, wide neck, vented, LaboPlast®
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0310-2056 0310-2054 0310-2055 0310-2051 0310-2052
BURK0310-2056EA 12.6 EUR
BURK0310-2056 BURK0310-2054 BURK0310-2055 BURK0310-2051 BURK0310-2052
Safety wash bottles, wide neck, vented, LaboPlast®
Flessen Spuitflessen
LDPE, wide-neck bottles.
  • With light protection
  • Safe filling due to wide-necked opening 39 mm neck diameter
  • Permanent print according to GHS with substance name in German, English, French and Spanish, GHS pictograms and CAS number
If the room temperature increases, or the sun shines into the lab, conventional wash bottles will drip because of the increased inner pressure. The venting valve ensures that the excess pressure can escape and dripping is prevented. With leakproof venting valve, contents cannot leak when bottle has fallen over as with washMore Product Information