Sample bags with closure, SteriBag StandUp
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5344-0400 5344-0600
BURK5344-0400EA 130 EUR
BURK5344-0400 BURK5344-0600
Sample bags with closure, SteriBag StandUp
Zakken Monsterzakken
SteriBag StandUp are sterile sample bags with a stand-up base. The special bottom unfolds as soon as the bag is filled. This makes SteriBag StandUp stable and able to stand on its own without a separate stand or support, leaving both your hands free to take the sample and fill the bag. The wire bracket at the closure keeps the bag open, making it easy and convenient to fill. Can be used for samples available as liquids, powders, solids and pastes; suitable for foods and can be frozen.
  • Space-saving, unbreakable, inexpensive and sterile replacement for