Splinter-proof titrating burettes, LaboPlast®
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9695-3025 9695-3050 9695-3015
612-5501EA 79.3 EUR
612-5501 612-5502 612-5500
Splinter-proof titrating burettes, LaboPlast®
Buretten
This titrating burette with splinter protection is a fast acting burette with Schellbach stripes based on Dr. Schilling's design with automatic zero point extraction and Bürkle dosing armature.
  • Burette with splinter-proof sheathing
  • Adjusted to Ex
  • Burette made of borosilicate glass 3.3
  • Shatter-proof, resistant to chemicals
  • Sturdy stand
  • Precise discharge jet, allowing very fine drops
  • Parts in contact with medium: PE, PP, natural rubber, borosilicate glass
The glass burette has a plastic sheathing which increases resistance to breaking and protects against splinters. The possibility of the burette breaking is much lower than with a conventional burette.
The Bürkle dosing armature allows for precise dosing and exact fine titration with the press of a button or micro-screw, and there is precise discharge through the discharge jet.