Gel permeation chromatography columns, Styragel® HR
Leverancier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WAT044231 WAT054405 WAT054415 WAT044229 WAT044228 WAT044237 WAT044222
WATEWAT044231EA 0 EUR
WATEWAT044231 WATEWAT054405 WATEWAT054415 WATEWAT044229 WATEWAT044228 WATEWAT044237 WATEWAT044222
Gel permeation chromatography columns, Styragel® HR
Kolommen voor chromatografie
Styragel® HR columns are high-resolution GPC columns packed with 5 µm particles and are ideal for characterising samples in the low to mid molecular range where high resolution is critical.