D-Glucose Monohydrate (Dextrose), cell culture tested
Leverancier: BIOWEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P5030-500GR P5030-1KG
P5030-500GREA 71.4 EUR
P5030-500GR P5030-1KG
D-Glucose Monohydrate (Dextrose), cell culture tested
Celcultuursupplementen en -additieven
D-glucose monohydrate (Dextrose) is a common natural sugar involved in processes such as energy production, glycosylation, and formation of glycans that provide structure to cells. It is involved in a detrimental process in cells called glycation. It is used as a supplement for cell culture and in numerous cellular processes.