ATP-Glo™ Bioluminometric cell viability assay
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30020-2 30020-1 30020-T
30020-2EA 570 EUR
30020-2 30020-1 30020-T
ATP-Glo™ Bioluminometric cell viability assay
Testen Cellulaire assays
ATP-Glo™ Bioluminometric Cell Viability Assay offers a highly sensitive assay for quantifying ATP. The homogeneous assay involves a single addition of ATP-Glo™ Detection Cocktail directly to cells in culture medium.
  • Highly sensitive, detect from a single cell to tens of thousands of cells
  • Excellent linearity over 6 orders of magnitude
  • Homogeneous, no-wash assay
  • Detect ATP amount or quantify number of live cells
ATP-Glo™ Bioluminometric Cell Viability Assay offers a highly sensitive assay for quantifying ATP. The homogeneous assay procedure involves simply adding the ATP-Glo™ detection cocktail directly to cells cultured in complete culture medium. It is not necessary to remove medium or wash cells before adding the reagent.
Kit Components: D-Luciferin, Firefly luciferase (recombinant, produced in E. coli), ATP-Glo™ assay buffer and 2 mm ATP standard.
This ATP detection kit takes advantage of firefly luciferase’s use of ATP to oxidize D-Luciferin and the resulting production of light in order to assess the amount of ATP available. Because ATP is an indicator of metabolically active cells, the number of viable cells can be assessed based on the amount of ATP present.
The ATP-Glo™ kit can be used to detect as little as a single cell or 0,01 picomole of ATP. The signal produced is linear within 6 orders of magnitude. By relating the amount of ATP to the number of viable cells, the assay has wide applications, ranging from the determination of viable cell numbers to cell proliferation to cell cytotoxicity.
Opgelet: ATP-Glo™ is a flash-type luminescence assay. The luminescence signal generated is stable for up to 1 minute. This assay is designed for individual sample detection by using a luminometer in a single sample format or a luminometer with an injector in 96-well plate format.